Assay Design



Gene Transcripts

Gene Symbol
map-2
Entrez Gene ID
178468
Gene Name
protein-coding
Gene Aliases
CELE_Y116A8A.9
Chromosome Location
Chr.IV: 16848154 - 16854924 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001268977.4 NP_001255906.1 3 - 4 530 55
NM_001268978.4 NP_001255907.1 4 - 5 560 55
NM_001372970.2 NP_001360158.1 3 - 3 488 55

View Full Product Details