Assay Design



Gene Transcripts

Gene Symbol
mel-46
Entrez Gene ID
178469
Gene Name
protein-coding
Gene Aliases
CELE_T06A10.1
Chromosome Location
Chr.IV: 16855044 - 16863449 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001268980.3 NP_001255909.1 2 - 3 375 102
NM_001372971.2 NP_001360155.1 2 - 3 375 102
GenBank mRNA EU051652.1 - 2 - 3 391 102

View Full Product Details