Assay Design



Gene Transcripts

Gene Symbol
mel-46
Entrez Gene ID
178469
Gene Name
protein-coding
Gene Aliases
CELE_T06A10.1
Chromosome Location
Chr.IV: 16855044 - 16863449 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001268980.3 NP_001255909.1 5 - 6 693 61
NM_001372971.2 NP_001360155.1 5 - 6 693 61
GenBank mRNA EU051652.1 - 5 - 6 709 61

View Full Product Details