Assay Design
Gene Transcripts
- Gene Symbol
- snr-1
- Entrez Gene ID
-
178483
- Gene Name
- Small nuclear ribonucleoprotein Sm D3
- Gene Aliases
-
CELE_Y116A8C.42
- Chromosome Location
-
Chr.IV: 17088058 - 17088946 on Build WBcel235
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_070626.6 |
NP_503027.1 |
2 - 3 |
205 |
118 |
View Full Product Details