Assay Design
Gene Transcripts
- Gene Symbol
- sfa-1
- Entrez Gene ID
-
178486
- Gene Name
- CCHC-type domain-containing protein
- Gene Aliases
-
CELE_Y116A8C.32
- Chromosome Location
-
Chr.IV: 17103810 - 17110982 on Build WBcel235
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_070632.6 |
NP_503033.1 |
4 - 5 |
733 |
82 |
GenBank mRNA |
AJ243905.1 |
- |
4 - 5 |
733 |
82 |
View Full Product Details