Assay Design
Gene Transcripts
- Gene Symbol
- srt-52
- Entrez Gene ID
-
189650
- Gene Name
- Serpentine Receptor, class T
- Gene Aliases
-
CELE_Y38C9B.2
- Chromosome Location
-
Chr.V: 49956 - 56046 on Build WBcel235
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_070728.2 |
NP_503129.2 |
4 - 5 |
837 |
155 |
View Full Product Details