Assay Design
Gene Transcripts
- Gene Symbol
- ttb-1
- Entrez Gene ID
-
178544
- Gene Name
- Transcription initiation factor IIB
- Gene Aliases
-
CELE_W03F9.5
- Chromosome Location
-
Chr.V: 156825 - 166866 on Build WBcel235
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_070745.5 |
NP_503146.1 |
2 - 3 |
388 |
79 |
View Full Product Details