Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001306844.3 | NP_001293773.1 | 9 - 10 | 1354 | 99 |
NM_001380293.1 | NP_001368400.1 | 8 - 9 | 1213 | 99 | |
NM_068657.7 | NP_501058.2 | 11 - 12 | 1612 | 99 | |
GenBank mRNA | AJ512334.1 | - | 10 - 11 | 1589 | 99 |