Assay Design
Gene Transcripts
- Gene Symbol
- rpn-1
- Entrez Gene ID
-
177455
- Gene Name
- 26S proteasome non-ATPase regulatory subunit 2
- Gene Aliases
-
CELE_T22D1.9
- Chromosome Location
-
Chr.IV: 6906370 - 6909913 on Build WBcel235
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_068663.5 |
NP_501064.1 |
3 - 4 |
1774 |
65 |
View Full Product Details