Assay Design
Gene Transcripts
- Gene Symbol
- ruvb-2
- Entrez Gene ID
-
177458
- Gene Name
- RuvB-like 2
- Gene Aliases
-
CELE_T22D1.10
- Chromosome Location
-
Chr.IV: 6917107 - 6920192 on Build WBcel235
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_068666.5 |
NP_501067.1 |
3 - 4 |
275 |
64 |
View Full Product Details