Assay Design



Gene Transcripts

Gene Symbol
srx-50
Entrez Gene ID
177478
Gene Name
protein-coding
Gene Aliases
CELE_Y43B11AL.2
Chromosome Location
Chr.IV: 7057977 - 7059911 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001380302.1 NP_001367449.1 6 - 7 801 83
NM_001380303.1 NP_001367448.1 6 - 7 782 83

View Full Product Details