Assay Design
Gene Transcripts
- Gene Symbol
- srh-80
- Entrez Gene ID
-
191874
- Gene Name
- Serpentine Receptor, class H
- Gene Aliases
-
CELE_DC2.1
- Chromosome Location
-
Chr.V: 219932 - 222776 on Build WBcel235
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_070756.1 |
NP_503157.1 |
1 - 2 |
210 |
163 |
View Full Product Details