Assay Design



Gene Transcripts

Gene Symbol
srh-80
Entrez Gene ID
191874
Gene Name
Serpentine Receptor, class H
Gene Aliases
CELE_DC2.1
Chromosome Location
Chr.V: 219932 - 222776 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_070756.1 NP_503157.1 2 - 3 362 122
NR_132473.1 2 - 3 465 122

View Full Product Details