Assay Design
Gene Transcripts
- Gene Symbol
- srj-37
- Entrez Gene ID
-
178579
- Gene Name
- Serpentine Receptor, class J
- Gene Aliases
-
CELE_F48G7.1
- Chromosome Location
-
Chr.V: 638628 - 639928 on Build WBcel235
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_070874.1 |
NP_503275.1 |
3 - 4 |
657 |
72 |
View Full Product Details