Assay Design
Gene Transcripts
- Gene Symbol
- srbc-40
- Entrez Gene ID
-
186776
- Gene Name
- protein-coding
- Gene Aliases
-
CELE_H24O09.1
- Chromosome Location
-
Chr.V: 715833 - 719831 on Build WBcel235
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_070903.2 |
NP_503304.2 |
1 - 2 |
91 |
116 |
View Full Product Details