Assay Design



Gene Transcripts

Gene Symbol
srh-2
Entrez Gene ID
178584
Gene Name
protein-coding
Gene Aliases
CELE_C05E4.14
Chromosome Location
Chr.V: 728312 - 734307 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001313113.1 NP_001300042.1 4 - 5 583 178
NM_001313116.1 NP_001300045.1 2 - 3 472 178
NM_001380592.1 NP_001367443.1 4 - 5 565 178
NM_001383271.1 5 - 6 766 178

View Full Product Details