Assay Design
Gene Transcripts
- Gene Symbol
- fmo-5
- Entrez Gene ID
-
178596
- Gene Name
- protein-coding
- Gene Aliases
-
CELE_H24K24.5
- Chromosome Location
-
Chr.V: 899304 - 903665 on Build WBcel235
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_070951.5 |
NP_503352.1 |
2 - 3 |
272 |
90 |
GenBank mRNA |
AJ582073.1 |
- |
2 - 3 |
186 |
90 |
View Full Product Details