Assay Design
Gene Transcripts
- Gene Symbol
- oac-31
- Entrez Gene ID
-
178643
- Gene Name
- NRF domain-containing protein
- Gene Aliases
-
CELE_F52F10.3
- Chromosome Location
-
Chr.V: 1538354 - 1541886 on Build WBcel235
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_071040.5 |
NP_503441.2 |
3 - 4 |
1252 |
58 |
View Full Product Details