Assay Design
Gene Transcripts
- Gene Symbol
- srh-5
- Entrez Gene ID
-
191842
- Gene Name
- Serpentine Receptor, class H
- Gene Aliases
-
CELE_K04F1.16
- Chromosome Location
-
Chr.V: 1685628 - 1686864 on Build WBcel235
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_071081.1 |
NP_503482.1 |
4 - 5 |
824 |
83 |
View Full Product Details