Assay Design
Gene Transcripts
- Gene Symbol
- srw-99
- Entrez Gene ID
-
189965
- Gene Name
- protein-coding
- Gene Aliases
-
CELE_Y46H3C.2
- Chromosome Location
-
Chr.V: 1708741 - 1710663 on Build WBcel235
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_071086.1 |
NP_503487.1 |
2 - 3 |
257 |
112 |
View Full Product Details