Assay Design



Gene Transcripts

Gene Symbol
ckb-4
Entrez Gene ID
184864
Gene Name
Choline Kinase B
Gene Aliases
CELE_F22F7.5
Chromosome Location
Chr.V: 2106398 - 2109434 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_071172.6 NP_503573.1 4 - 5 642 76
NR_132528.1 3 - 4 490 76

View Full Product Details