Assay Design
Gene Transcripts
- Gene Symbol
- srbc-23
- Entrez Gene ID
-
183490
- Gene Name
- Serpentine Receptor, class BC (class B-like)
- Gene Aliases
-
CELE_C45H4.9
- Chromosome Location
-
Chr.V: 2153574 - 2155799 on Build WBcel235
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_071182.3 |
NP_503583.2 |
1 - 2 |
109 |
163 |
View Full Product Details