Assay Design



Gene Transcripts

Gene Symbol
cyp-33C6
Entrez Gene ID
185591
Gene Name
protein-coding
Gene Aliases
CELE_F41B5.7
Chromosome Location
Chr.V: 2247254 - 2249410 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001316799.2 NP_001303728.1 1 - 2 272 109
NM_001380616.1 NP_001368585.1 1 - 2 95 109

View Full Product Details