Assay Design
Gene Transcripts
- Gene Symbol
- cyp-33C7
- Entrez Gene ID
-
185589
- Gene Name
- CYtochrome P450 family
- Gene Aliases
-
CELE_F41B5.2
- Chromosome Location
-
Chr.V: 2250540 - 2252591 on Build WBcel235
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_071217.3 |
NP_503618.2 |
1 - 2 |
210 |
62 |
View Full Product Details