Assay Design



Gene Transcripts

Gene Symbol
ketn-1
Entrez Gene ID
178740
Gene Name
protein-coding
Gene Aliases
CELE_F54E2.3
Chromosome Location
Chr.V: 2781939 - 2807860 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001307737.5 NP_001294666.1 25 - 26 14536 103
NM_001307738.4 NP_001294667.1 24 - 25 14413 103
NM_001307739.4 NP_001294668.1 27 - 28 14758 103
NM_001307740.4 NP_001294669.1 26 - 27 14635 103
NM_001316807.3 NP_001303736.1 12 - 13 10060 103
NM_001316808.3 NP_001303737.1 9 - 10 9649 103
NM_001377590.2 NP_001364781.1 16 - 17 12496 103
NM_001377591.2 NP_001364828.1 18 - 19 12718 103
NM_001377593.1 NP_001364783.1 17 - 18 12619 103
NM_001377594.2 NP_001364827.1 19 - 20 12841 103
GenBank mRNA AY819766.1 - 20 - 21 13406 103

View Full Product Details