Assay Design
Gene Transcripts
- Gene Symbol
- srj-57
- Entrez Gene ID
-
191954
- Gene Name
- Serpentine Receptor, class J
- Gene Aliases
-
CELE_T03D3.14
- Chromosome Location
-
Chr.V: 2854393 - 2855573 on Build WBcel235
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_071371.1 |
NP_503772.1 |
2 - 3 |
556 |
81 |
View Full Product Details