Assay Design
Gene Transcripts
- Gene Symbol
- sre-24
- Entrez Gene ID
-
185415
- Gene Name
- Serpentine Receptor, class E (epsilon)
- Gene Aliases
-
CELE_F37B4.9
- Chromosome Location
-
Chr.V: 2884711 - 2886425 on Build WBcel235
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_071383.1 |
NP_503784.1 |
2 - 3 |
604 |
90 |
View Full Product Details