Assay Design
Gene Transcripts
- Gene Symbol
- srt-8
- Entrez Gene ID
-
183692
- Gene Name
- Serpentine Receptor, class T
- Gene Aliases
-
CELE_C50H11.2
- Chromosome Location
-
Chr.V: 3086560 - 3087986 on Build WBcel235
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_071443.1 |
NP_503844.1 |
3 - 4 |
717 |
133 |
View Full Product Details