Assay Design
Gene Transcripts
- Gene Symbol
- cyp-33C9
- Entrez Gene ID
-
178752
- Gene Name
- CYtochrome P450 family
- Gene Aliases
-
CELE_C50H11.15
- Chromosome Location
-
Chr.V: 3091812 - 3094561 on Build WBcel235
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_071445.5 |
NP_503846.1 |
4 - 5 |
1289 |
81 |
View Full Product Details