Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001373012.2 | NP_001360546.1 | 5 - 6 | 926 | 64 |
NM_071481.5 | NP_503882.2 | 5 - 6 | 933 | 64 | |
NR_134544.1 | 4 - 5 | 1028 | 64 | ||
NR_134545.1 | 4 - 5 | 617 | 64 | ||
NR_134546.1 | 2 - 3 | 371 | 64 | ||
GenBank mRNA | U07628.1 | - | 5 - 6 | 931 | 64 |