Assay Design
Gene Transcripts
- Gene Symbol
- srj-8
- Entrez Gene ID
-
189015
- Gene Name
- Serpentine Receptor, class J
- Gene Aliases
-
CELE_T28A11.9
- Chromosome Location
-
Chr.V: 3247815 - 3249140 on Build WBcel235
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_071490.1 |
NP_503891.1 |
1 - 2 |
190 |
71 |
View Full Product Details