Assay Design



Gene Transcripts

Gene Symbol
srj-9
Entrez Gene ID
191934
Gene Name
protein-coding
Gene Aliases
CELE_T20C4.1
Chromosome Location
Chr.V: 3431940 - 3433751 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001322515.1 NP_001309651.1 1 - 2 194 123
NM_071562.2 NP_503963.1 1 - 2 513 123

View Full Product Details