Assay Design
Gene Transcripts
- Gene Symbol
- cfz-2
- Entrez Gene ID
-
178768
- Gene Name
- Frizzled-2
- Gene Aliases
-
CELE_F27E11.3
- Chromosome Location
-
Chr.V: 3441504 - 3444879 on Build WBcel235
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_071564.7 |
NP_503965.2 |
3 - 4 |
1608 |
77 |
GenBank mRNA |
DQ178242.1 |
- |
3 - 4 |
1606 |
77 |
View Full Product Details