Assay Design
Gene Transcripts
- Gene Symbol
- srx-66
- Entrez Gene ID
-
187097
- Gene Name
- G_PROTEIN_RECEP_F1_2 domain-containing protein
- Gene Aliases
-
CELE_K07C6.9
- Chromosome Location
-
Chr.V: 3919589 - 3920825 on Build WBcel235
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_071684.4 |
NP_504085.2 |
1 - 2 |
196 |
128 |
View Full Product Details