Assay Design



Gene Transcripts

Gene Symbol
srx-64
Entrez Gene ID
187095
Gene Name
G_PROTEIN_RECEP_F1_2 domain-containing protein
Gene Aliases
CELE_K07C6.7
Chromosome Location
Chr.V: 3929399 - 3930678 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001330820.1 NP_001317762.1 2 - 3 208 101

View Full Product Details