Assay Design
Gene Transcripts
- Gene Symbol
- phat-5
- Entrez Gene ID
-
178811
- Gene Name
- PHAryngeal gland Toxin-related
- Gene Aliases
-
CELE_T05B4.11
- Chromosome Location
-
Chr.V: 4121489 - 4122517 on Build WBcel235
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_071747.6 |
NP_504148.1 |
1 - 2 |
155 |
83 |
View Full Product Details