Assay Design
Gene Transcripts
- Gene Symbol
- srt-14
- Entrez Gene ID
-
191088
- Gene Name
- Serpentine Receptor, class T
- Gene Aliases
-
CELE_ZC142.2
- Chromosome Location
-
Chr.V: 4253502 - 4256574 on Build WBcel235
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_071773.2 |
NP_504174.2 |
1 - 2 |
93 |
107 |
View Full Product Details