Assay Design
Gene Transcripts
- Gene Symbol
- srt-38
- Entrez Gene ID
-
185927
- Gene Name
- Serpentine Receptor, class T
- Gene Aliases
-
CELE_F47D2.6
- Chromosome Location
-
Chr.V: 4269455 - 4271358 on Build WBcel235
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_071777.2 |
NP_504178.1 |
3 - 4 |
711 |
75 |
View Full Product Details