Assay Design
Gene Transcripts
- Gene Symbol
- oac-12
- Entrez Gene ID
-
183958
- Gene Name
- Acyl_transf_3 domain-containing protein
- Gene Aliases
-
CELE_D2063.2
- Chromosome Location
-
Chr.V: 4329039 - 4332550 on Build WBcel235
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_071795.1 |
NP_504196.1 |
5 - 6 |
1134 |
89 |
View Full Product Details