Assay Design
Gene Transcripts
- Gene Symbol
- srx-77
- Entrez Gene ID
-
186920
- Gene Name
- 7TM_GPCR_Srx domain-containing protein
- Gene Aliases
-
CELE_K03B4.5
- Chromosome Location
-
Chr.V: 4666745 - 4668736 on Build WBcel235
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_071856.3 |
NP_504257.3 |
3 - 4 |
469 |
129 |
View Full Product Details