Assay Design



Gene Transcripts

Gene Symbol
ugt-9
Entrez Gene ID
178883
Gene Name
protein-coding
Gene Aliases
CELE_T19H12.1
Chromosome Location
Chr.V: 4896028 - 4897969 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001129547.4 NP_001123019.1 3 - 4 375 120
NM_071911.6 NP_504312.1 3 - 4 400 120

View Full Product Details