Assay Design
Gene Transcripts
- Gene Symbol
- srr-6
- Entrez Gene ID
-
182569
- Gene Name
- Serpentine Receptor, class R
- Gene Aliases
-
CELE_C13D9.1
- Chromosome Location
-
Chr.V: 5012803 - 5015219 on Build WBcel235
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_071945.3 |
NP_504346.1 |
3 - 4 |
1061 |
126 |
View Full Product Details