Assay Design
Gene Transcripts
- Gene Symbol
- srab-1
- Entrez Gene ID
-
182220
- Gene Name
- Serpentine Receptor, class AB (class A-like)
- Gene Aliases
-
CELE_C04F5.4
- Chromosome Location
-
Chr.V: 5100098 - 5102117 on Build WBcel235
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_071965.1 |
NP_504366.1 |
2 - 3 |
445 |
116 |
View Full Product Details