Assay Design
Gene Transcripts
- Gene Symbol
- srab-2
- Entrez Gene ID
-
182221
- Gene Name
- Serpentine Receptor, class AB (class A-like)
- Gene Aliases
-
CELE_C04F5.5
- Chromosome Location
-
Chr.V: 5103042 - 5105034 on Build WBcel235
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_071966.1 |
NP_504367.1 |
4 - 5 |
636 |
120 |
View Full Product Details