Assay Design
Gene Transcripts
- Gene Symbol
- cft-1
- Entrez Gene ID
-
178939
- Gene Name
- Cystic Fibrosis Transmembrane conductance regulator homolog
- Gene Aliases
-
CELE_C18C4.2
- Chromosome Location
-
Chr.V: 5538800 - 5545789 on Build WBcel235
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_072062.5 |
NP_504463.1 |
15 - 16 |
2788 |
152 |
View Full Product Details