Assay Design
Gene Transcripts
- Gene Symbol
- ugt-48
- Entrez Gene ID
-
178940
- Gene Name
- Putative UDP-glucuronosyltransferase ugt-48
- Gene Aliases
-
CELE_C18C4.3
- Chromosome Location
-
Chr.V: 5545650 - 5548322 on Build WBcel235
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_072063.7 |
NP_504464.2 |
4 - 5 |
615 |
74 |
View Full Product Details