Assay Design
Gene Transcripts
- Gene Symbol
- mut-14
- Entrez Gene ID
-
178952
- Gene Name
- MUTator
- Gene Aliases
-
CELE_C14C11.6
- Chromosome Location
-
Chr.V: 5658698 - 5660704 on Build WBcel235
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_072089.5 |
NP_504490.1 |
1 - 2 |
136 |
73 |
View Full Product Details