Assay Design
Gene Transcripts
- Gene Symbol
- fut-8
- Entrez Gene ID
-
178985
- Gene Name
- Alpha-(1,6)-fucosyltransferase
- Gene Aliases
-
CELE_C10F3.6
- Chromosome Location
-
Chr.V: 5986990 - 5990252 on Build WBcel235
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_072154.8 |
NP_504555.2 |
7 - 8 |
1390 |
121 |
GenBank mRNA |
AJ512486.1 |
- |
7 - 8 |
1368 |
121 |
View Full Product Details