Assay Design



Gene Transcripts

Gene Symbol
acl-8
Entrez Gene ID
179031
Gene Name
PlsC domain-containing protein
Gene Aliases
CELE_T05H4.1
Chromosome Location
Chr.V: 6450870 - 6453119 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001361761.3 NP_001348648.1 2 - 3 652 90

View Full Product Details