Assay Design
Gene Transcripts
- Gene Symbol
- ubh-1
- Entrez Gene ID
-
179039
- Gene Name
- Ubiquitin carboxyl-terminal hydrolase
- Gene Aliases
-
CELE_F46E10.8
- Chromosome Location
-
Chr.V: 6508928 - 6511409 on Build WBcel235
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_072253.5 |
NP_504654.1 |
4 - 5 |
337 |
65 |
View Full Product Details